CO1H324A6E83 CombiPhos Catalysts Inc

4,4,5,5-tetramethyl-2-(4-methyl-3-(tetrahydro-2H-pyran-3-yl)phenyl)-1,3,2-dioxaborolane

  • Cas Number : 2222996-57-6
  • Molecular Weight : 302.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart