CO1H324A6EA0 CombiPhos Catalysts Inc

4-(3-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine

  • Cas Number : 2223053-57-2
  • Molecular Weight : 317.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart