CO1H324A6EC5 CombiPhos Catalysts Inc

methyl 5-cyclopropyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • Cas Number : 2223032-46-8
  • Molecular Weight : 302.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart