CO1H324A6ED4 CombiPhos Catalysts Inc

3-methyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)piperidine

  • Cas Number : 2223008-89-5
  • Molecular Weight : 369.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart