CO1H324A6ED7 CombiPhos Catalysts Inc

4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)morpholine

  • Cas Number : 651014-91-4
  • Molecular Weight : 357.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart