CO1H324A6EDA CombiPhos Catalysts Inc

1-[3-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-5-TRIFLUOROMETHYLPHENYL]PIPERIDIN-2-ONE

  • Cas Number : 1192052-35-9
  • Molecular Weight : 369.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart