CO1H324A6EDE CombiPhos Catalysts Inc

1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)-1H-imidazole

  • Cas Number : 2223050-09-5
  • Molecular Weight : 338.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart