CO1H324A6EDF CombiPhos Catalysts Inc

2-methyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)-1H-imidazole

  • Cas Number : 2223030-83-7
  • Molecular Weight : 352.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart