CO1H324A6EE3 CombiPhos Catalysts Inc

6-(Cyclopropyl)imidazo[1,2-a]pyrazine-8-boronic acid pinacol ester

  • Cas Number : 2223041-14-1
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart