CO1H324A6EE7 CombiPhos Catalysts Inc

Imidazo[1,2-a]pyrazine-6-boronic acid pinacol ester

  • Cas Number : 1445860-58-1
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart