CO1H324A6EE8 CombiPhos Catalysts Inc

4-methyl-1-(3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole

  • Cas Number : 2223053-72-1
  • Molecular Weight : 298.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart