CO1H324A6EED CombiPhos Catalysts Inc

1-[3-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-5-METHYLPHENYL]PYRROLIDIN-2-ONE

  • Cas Number : 2223033-53-0
  • Molecular Weight : 301.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart