CO1H324A6EF3 CombiPhos Catalysts Inc

1-[3-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-5-METHYLPHENYL]PIPERIDIN-2-ONE

  • Cas Number : 2223030-78-0
  • Molecular Weight : 315.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart