CO1H324A6EFD CombiPhos Catalysts Inc

6-(Piperazin-1-yl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 1309981-38-1
  • Molecular Weight : 289.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart