CO1H324A6F15 CombiPhos Catalysts Inc

3-Bromo-5-(3-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2222997-59-1
  • Molecular Weight : 380.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart