CO1H324A6F16 CombiPhos Catalysts Inc

3-Bromo-5-(4-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223054-89-3
  • Molecular Weight : 380.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart