CO1H324A6F17 CombiPhos Catalysts Inc

4-(3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine

  • Cas Number : 1893410-04-2
  • Molecular Weight : 368.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart