CO1H324A6F1B CombiPhos Catalysts Inc

3-Fluoro-4-(1H-pyrrol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223031-01-2
  • Molecular Weight : 287.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart