CO1H324A6F1C CombiPhos Catalysts Inc

3-Fluoro-4-(imidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223053-40-3
  • Molecular Weight : 288.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart