CO1H324A6F1D CombiPhos Catalysts Inc

3-Fluoro-4-(2-methylimidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223042-23-5
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart