CO1H324A6F25 CombiPhos Catalysts Inc

3-Fluoro-4-(4-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223045-90-5
  • Molecular Weight : 319.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart