CO1H324A6F27 CombiPhos Catalysts Inc

1-[4-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-2-FLUOROPHENYL]PIPERIDIN-2-ONE

  • Cas Number : 2223042-39-3
  • Molecular Weight : 319.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart