CO1H324A6F2E CombiPhos Catalysts Inc

tert-butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenyl)piperazine-1-carboxylate

  • Cas Number : 1796562-08-7
  • Molecular Weight : 456.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart