CO1H324A6F2F CombiPhos Catalysts Inc

3-(Trifluoromethyl)-4-(morpholino)phenylboronic acid pinacol ester

  • Cas Number : 1373522-71-4
  • Molecular Weight : 357.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart