CO1H324A6F30 CombiPhos Catalysts Inc

1-[4-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-2-TRIFLUOROMETHYLPHENYL]PIPERIDIN-2-ONE

  • Cas Number : 2223054-91-7
  • Molecular Weight : 369.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart