CO1H324A6F45 CombiPhos Catalysts Inc

3-Methyl-4-(4-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223035-02-5
  • Molecular Weight : 315.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart