CO1H324A6F4A CombiPhos Catalysts Inc

3-Methyl-4-(N-methylpiperazin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 1310704-51-8
  • Molecular Weight : 316.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart