CO1H324A6F56 CombiPhos Catalysts Inc

2-Methyl-4-(imidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223040-14-8
  • Molecular Weight : 284.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart