CO1H324A6F58 CombiPhos Catalysts Inc

2-Methyl-4-(4-methylimidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223033-37-0
  • Molecular Weight : 298.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart