CO1H324A6F5F CombiPhos Catalysts Inc

3-Fluoro-5-(1H-pyridin-2-one)phenylboronic acid pinacol ester

  • Cas Number : 2223053-97-0
  • Molecular Weight : 315.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart