CO1H324A6F6A CombiPhos Catalysts Inc

2-Methoxy-4-(2-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223004-65-5
  • Molecular Weight : 331.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart