CO1H324A6F70 CombiPhos Catalysts Inc

4-(Cyclopropyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223041-50-5
  • Molecular Weight : 250.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart