CO1H324A6F73 CombiPhos Catalysts Inc

2-Methoxy-4-(1H-pyridin-2-one)phenylboronic acid pinacol ester

  • Cas Number : 2223033-65-4
  • Molecular Weight : 327.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart