CO1H324A6F79 CombiPhos Catalysts Inc

1-[4-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-3-BROMOPHENYL]PIPERIDIN-2-ONE

  • Cas Number : 2223040-18-2
  • Molecular Weight : 380.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart