CO1H324A6F7F CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(N-methylpiperazin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223041-59-4
  • Molecular Weight : 370.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart