CO1H324A6F82 CombiPhos Catalysts Inc

2-Fluoro-4-(N-methylpiperazin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2133421-20-0
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart