CO1H324A6F89 CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(morpholino)phenylboronic acid pinacol ester

  • Cas Number : 2223035-67-2
  • Molecular Weight : 357.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart