CO1H324A6F93 CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(3-methylpiperidin-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2222996-39-4
  • Molecular Weight : 369.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart