CO1H324A6F9E CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(piperidino)phenylboronic acid pinacol ester

  • Cas Number : 2223037-13-4
  • Molecular Weight : 355.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart