CO1H324A6FA1 CombiPhos Catalysts Inc

2-Fluoro-4-(piperidino)phenylboronic acid pinacol ester

  • Cas Number : 2223032-11-7
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart