CO1H324A6FA2 CombiPhos Catalysts Inc

1-[4-(TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-3-BROMOPHENYL]PYRROLIDIN-2-ONE

  • Cas Number : 2223046-11-3
  • Molecular Weight : 366.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart