CO1H324A6FA7 CombiPhos Catalysts Inc

2-Bromo-4-(pyrrolidino)phenylboronic acid pinacol ester

  • Cas Number : 2223033-84-7
  • Molecular Weight : 352.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart