CO1H324A6FA8 CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(pyrrolidino)phenylboronic acid pinacol ester

  • Cas Number : 2223031-45-4
  • Molecular Weight : 341.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart