CO1H324A6FB6 CombiPhos Catalysts Inc

2-Bromo-4-(imidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223033-40-5
  • Molecular Weight : 349.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart