CO1H324A6FC2 CombiPhos Catalysts Inc

2-Chloro-4-(1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223035-75-2
  • Molecular Weight : 304.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart