CO1H324A6FC4 CombiPhos Catalysts Inc

2-Fluoro-4-(1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 1688637-43-5
  • Molecular Weight : 288.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart