CO1H324A6FCB CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(4-methyl-1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223054-16-6
  • Molecular Weight : 352.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart