CO1H324A6FCD CombiPhos Catalysts Inc

2-Cyano-4-(4-methyl-1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223032-98-0
  • Molecular Weight : 309.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart