CO1H324A6FD0 CombiPhos Catalysts Inc

2-Methyl-4-cyano-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223041-84-5
  • Molecular Weight : 283.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart