CO1H324A6FD4 CombiPhos Catalysts Inc

2-Methyl-3-cyano-5-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223040-40-0
  • Molecular Weight : 283.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart